MilliporeSigma
  • Aspergillus fumigatus CalA binds to integrin α5β1 and mediates host cell invasion.
Aspergillus fumigatus CalA binds to integrin α5β1 and mediates host cell invasion.
Nature microbiology (2016-11-15)
Hong Liu, Mark J Lee, Norma V Solis, Quynh T Phan, Marc Swidergall, Benjamin Ralph, Ashraf S Ibrahim, Donald C Sheppard, Scott G Filler
ABSTRACT
Aspergillus fumigatus is an opportunistic fungal pathogen that invades pulmonary epithelial cells and vascular endothelial cells by inducing its own endocytosis, but the mechanism by which this process occurs is poorly understood. Here, we show that the thaumatin-like protein CalA is expressed on the surface of the A. fumigatus cell wall, where it mediates invasion of epithelial and endothelial cells. CalA induces endocytosis in part by interacting with integrin α5β1 on host cells. In corticosteroid-treated mice, a ΔcalA deletion mutant has significantly attenuated virulence relative to the wild-type strain, as manifested by prolonged survival, reduced pulmonary fungal burden and decreased pulmonary invasion. Pretreatment with an anti-CalA antibody improves survival of mice with invasive pulmonary aspergillosis, demonstrating the potential of CalA as an immunotherapeutic target. Thus, A. fumigatus CalA is an invasin that interacts with integrin α5β1 on host cells, induces endocytosis and enhances virulence.
Sigma-Aldrich
Anti-Integrin α3 Antibody, clone P1B5, azide free, clone P1B5, Chemicon®, from mouse