MilliporeSigma
  • Gα12 facilitates shortening in human airway smooth muscle by modulating phosphoinositide 3-kinase-mediated activation in a RhoA-dependent manner.
Gα12 facilitates shortening in human airway smooth muscle by modulating phosphoinositide 3-kinase-mediated activation in a RhoA-dependent manner.
British journal of pharmacology (2017-09-19)
Edwin J Yoo, Gaoyuan Cao, Cynthia J Koziol-White, Christie A Ojiaku, Krishna Sunder, Joseph A Jude, James V Michael, Hong Lam, Ivan Pushkarsky, Robert Damoiseaux, Dino Di Carlo, Kwangmi Ahn, Steven S An, Raymond B Penn, Reynold A Panettieri
ABSTRACT
PI3K-dependent activation of Rho kinase (ROCK) is necessary for agonist-induced human airway smooth muscle cell (HASMC) contraction, and inhibition of PI3K promotes bronchodilation of human small airways. The mechanisms driving agonist-mediated PI3K/ROCK axis activation, however, remain unclear. Given that G12 family proteins activate ROCK pathways in other cell types, their role in M3 muscarinic acetylcholine receptor-stimulated PI3K/ROCK activation and contraction was examined. Gα12 coupling was evaluated using co-immunoprecipitation and serum response element (SRE)-luciferase reporter assays. siRNA and pharmacological approaches, as well as overexpression of a regulator of G-protein signaling (RGS) proteins were applied in HASMCs. Phosphorylation levels of Akt, myosin phosphatase targeting subunit-1 (MYPT1), and myosin light chain-20 (MLC) were measured. Contraction and shortening were evaluated using magnetic twisting cytometry (MTC) and micro-pattern deformation, respectively. Human precision-cut lung slices (hPCLS) were utilized to evaluate bronchoconstriction. Knockdown of M3 receptors or Gα12 attenuated activation of Akt, MYPT1, and MLC phosphorylation. Gα12 coimmunoprecipitated with M3 receptors, and p115RhoGEF-RGS overexpression inhibited carbachol-mediated induction of SRE-luciferase reporter. p115RhoGEF-RGS overexpression inhibited carbachol-induced activation of Akt, HASMC contraction, and shortening. Moreover, inhibition of RhoA blunted activation of PI3K. Lastly, RhoA inhibitors induced dilation of hPCLS. Gα12 plays a crucial role in HASMC contraction via RhoA-dependent activation of the PI3K/ROCK axis. Inhibition of RhoA activation induces bronchodilation in hPCLS, and targeting Gα12 signaling may elucidate novel therapeutic targets in asthma. These findings provide alternative approaches to the clinical management of airway obstruction in asthma.
Sigma-Aldrich
Anti-MLC-2 Antibody, clone 19D3.1, clone 19D3.1, from mouse