MilliporeSigma
  • Nitric Oxide-induced Activation of the Type 1 Ryanodine Receptor Is Critical for Epileptic Seizure-induced Neuronal Cell Death.
Nitric Oxide-induced Activation of the Type 1 Ryanodine Receptor Is Critical for Epileptic Seizure-induced Neuronal Cell Death.
EBioMedicine (2016-08-22)
Yoshinori Mikami, Kazunori Kanemaru, Yohei Okubo, Takuya Nakaune, Junji Suzuki, Kazuki Shibata, Hiroki Sugiyama, Ryuta Koyama, Takashi Murayama, Akihiro Ito, Toshiko Yamazawa, Yuji Ikegaya, Takashi Sakurai, Nobuhito Saito, Sho Kakizawa, Masamitsu Iino
ABSTRACT
Status epilepticus (SE) is a life-threatening emergency that can cause neurodegeneration with debilitating neurological disorders. However, the mechanism by which convulsive SE results in neurodegeneration is not fully understood. It has been shown that epileptic seizures produce markedly increased levels of nitric oxide (NO) in the brain, and that NO induces Ca2+ release from the endoplasmic reticulum via the type 1 ryanodine receptor (RyR1), which occurs through S-nitrosylation of the intracellular Ca2+ release channel. Here, we show that through genetic silencing of NO-induced activation of the RyR1 intracellular Ca2+ release channel, neurons were rescued from seizure-dependent cell death. Furthermore, dantrolene, an inhibitor of RyR1, was protective against neurodegeneration caused by SE. These results demonstrate that NO-induced Ca2+ release via RyR is involved in SE-induced neurodegeneration, and provide a rationale for the use of RyR1 inhibitors for the prevention of brain damage following SE.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Ryanodine Receptor 1 Antibody, serum, Chemicon®