MilliporeSigma
  • The ANK repeats of Notch-4/Int3 activate NF-κB canonical pathway in the absence of Rbpj and causes mammary tumorigenesis.
The ANK repeats of Notch-4/Int3 activate NF-κB canonical pathway in the absence of Rbpj and causes mammary tumorigenesis.
Scientific reports (2017-10-24)
Ahmed Raafat, Sharon Bargo, David McCurdy, Robert Callahan
ABSTRACT
Transgenic mice expressing the Notch-4 intracellular domain (designated Int3) in the mammary gland have two phenotypes exhibited with 100% penetrance: arrest of mammary alveolar/lobular development and mammary tumorigenesis. Notch-4 signaling is mediated primarily through the interaction of Int3 with the transcription repressor/activator Rbpj. Interestingly, WAP-Int3/Rbpj knockout mice have normal mammary gland development but still developed mammary tumors with a slightly longer latency than the WAP-Int3 mice. Thus, Notch-induced mammary tumor development is Rbpj-independent. Here, we show that Int3 activates NF-κB in HC11 cells in absence of Rbpj through an association with the IKK signalosome. Int3 induced the canonical NF-κB activity and P50 phosphorylation in HC11 cells without altering the NF-κB2 pathway. The minimal domain within the Int3 protein required to activate NF-κB consists of the CDC10/Ankyrin (ANK) repeats domain. Treatment of WAP-Int3 tumor bearing mice with an IKK inhibitor resulted in tumor regression. In a soft agar assay, treatment of HC11-Int3 cells with P50-siRNA caused a significant decrease in colony formation. In addition, Wap-Int3/P50 knockout mice did not develop mammary tumors. This data indicates that the activation of NF-κB canonical signaling by Notch-4/Int3 is ANK repeats dependent, Rbpj-independent, and is mediated by IKK activation and P50 phosphorylation causing mammary tumorigenesis.
Sigma-Aldrich
Anti-Notch 4 Antibody, Upstate®, from rabbit