MilliporeSigma
[2]Rotaxane Formation by Transition State Stabilization.
Journal of the American Chemical Society (2017-06-18)
Guillaume De Bo, Guillaume Dolphijn, Charlie T McTernan, David A Leigh
ABSTRACT
We report on the synthesis of [2]rotaxanes driven by stabilization of the axle-forming transition state. A bifunctional macrocycle, with hydrogen bond donors at one end and acceptors at the other, is used to stabilize the charges that develop during the addition of a primary amine to a cyclic sulfate.