MilliporeSigma
  • Retargeting of human T cells to tumor-associated MUC1: the evolution of a chimeric antigen receptor.
Retargeting of human T cells to tumor-associated MUC1: the evolution of a chimeric antigen receptor.
Journal of immunology (Baltimore, Md. : 1950) (2008-03-21)
Scott Wilkie, Gianfranco Picco, Julie Foster, David M Davies, Sylvain Julien, Lucienne Cooper, Sefina Arif, Stephen J Mather, Joyce Taylor-Papadimitriou, Joy M Burchell, John Maher
ABSTRACT
MUC1 is a highly attractive immunotherapeutic target owing to increased expression, altered glycosylation, and loss of polarity in >80% of human cancers. To exploit this, we have constructed a panel of chimeric Ag receptors (CAR) that bind selectively to tumor-associated MUC1. Two parameters proved crucial in optimizing the CAR ectodomain. First, we observed that the binding of CAR-grafted T cells to anchored MUC1 is subject to steric hindrance, independent of glycosylation status. This was overcome by insertion of the flexible and elongated hinge found in immunoglobulins of the IgD isotype. Second, CAR function was highly dependent upon strong binding capacity across a broad range of tumor-associated MUC1 glycoforms. This was realized by using an Ab-derived single-chain variable fragment (scFv) cloned from the HMFG2 hybridoma. To optimize CAR signaling, tripartite endodomains were constructed. Ultimately, this iterative design process yielded a potent receptor termed HOX that contains a fused CD28/OX40/CD3zeta endodomain. HOX-expressing T cells proliferate vigorously upon repeated encounter with soluble or membrane-associated MUC1, mediate production of proinflammatory cytokines (IFN-gamma and IL-17), and elicit brisk killing of MUC1(+) tumor cells. To test function in vivo, a tumor xenograft model was derived using MDA-MB-435 cells engineered to coexpress MUC1 and luciferase. Mice bearing an established tumor were treated i.p. with a single dose of engineered T cells. Compared with control mice, this treatment resulted in a significant delay in tumor growth as measured by serial bioluminescence imaging. Together, these data demonstrate for the first time that the near-ubiquitous MUC1 tumor Ag can be targeted using CAR-grafted T cells.