MilliporeSigma
  • Novel Mutation in FLNC (Filamin C) Causes Familial Restrictive Cardiomyopathy.
Novel Mutation in FLNC (Filamin C) Causes Familial Restrictive Cardiomyopathy.
Circulation. Cardiovascular genetics (2017-12-08)
Nathan R Tucker, Micheal A McLellan, Dongjian Hu, Jiangchuan Ye, Victoria A Parsons, Robert W Mills, Sebastian Clauss, Elena Dolmatova, Marisa A Shea, David J Milan, Nandita S Scott, Mark Lindsay, Steven A Lubitz, Ibrahim J Domian, James R Stone, Honghuang Lin, Patrick T Ellinor
ABSTRACT
Restrictive cardiomyopathy (RCM) is a rare cardiomyopathy characterized by impaired diastolic ventricular function resulting in a poor clinical prognosis. Rarely, heritable forms of RCM have been reported, and mutations underlying RCM have been identified in genes that govern the contractile function of the cardiomyocytes. We evaluated 8 family members across 4 generations by history, physical examination, electrocardiography, and echocardiography. Affected individuals presented with a pleitropic syndrome of progressive RCM, atrioventricular septal defects, and a high prevalence of atrial fibrillation. Exome sequencing of 5 affected members identified a single novel missense variant in a highly conserved residue of FLNC (filamin C; p.V2297M). FLNC encodes filamin C-a protein that acts as both a scaffold for the assembly and organization of the central contractile unit of striated muscle and also as a mechanosensitive signaling molecule during cell migration and shear stress. Immunohistochemical analysis of FLNC localization in cardiac tissue from an affected family member revealed a diminished localization at the z disk, whereas traditional localization at the intercalated disk was preserved. Stem cell-derived cardiomyocytes mutated to carry the effect allele had diminished contractile activity when compared with controls. We have identified a novel variant in FLNC as pathogenic variant for familial RCM-a finding that further expands on the genetic basis of this rare and morbid cardiomyopathy.
Sigma-Aldrich
Anti-FLNC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution