MilliporeSigma
  • Reduced Cbl phosphorylation and degradation of the zeta-chain of the T-cell receptor/CD3 complex in T cells with low Lck levels.
Reduced Cbl phosphorylation and degradation of the zeta-chain of the T-cell receptor/CD3 complex in T cells with low Lck levels.
European journal of immunology (2008-09-16)
Trond Methi, Torunn Berge, Knut Martin Torgersen, Kjetil Taskén
ABSTRACT
T cells with short interfering RNA-mediated Lck-knockdown (kd) display paradoxical hyper-responsiveness upon TCR ligation. We have previously reported a possible mechanism for T-cell activation in cells with low levels of Lck depending on Grb2-SOS1 recruitment to the zeta-chain of TCR/CD3 (Methi et al., Eur. J. Immunol. 2007, 37: 2539-2548). Here, we show that short interfering RNA-mediated targeting of Lck caused a dramatic reduction in c-Cbl phosphorylation and a general reduction in protein ubiquitination after TCR stimulation. Specifically, this resulted in reduced ubiquitination of the zeta-chain, yet internalization of TCR/CD3 appeared to be normal after receptor engagement. However, zeta-chain levels were elevated in Lck-kd cells, and confocal microscopy revealed reduced colocalization of CD3-containing vesicles with endosomal and lysosomal compartments. We hypothesize that prolonged stability of internalized T-cell receptor complex may result in extended signaling in T cells with low Lck levels.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, recombinant clone 4G10®, clone 4G10®, Upstate®, from mouse