MilliporeSigma
  • Doxorubicin plus the IGF-1R antibody cixutumumab in soft tissue sarcoma: a phase I study using the TITE-CRM model.
Doxorubicin plus the IGF-1R antibody cixutumumab in soft tissue sarcoma: a phase I study using the TITE-CRM model.
Annals of oncology : official journal of the European Society for Medical Oncology (2015-04-11)
R Chugh, K A Griffith, E J Davis, D G Thomas, J D Zavala, G Metko, B Brockstein, S D Undevia, W M Stadler, S M Schuetze
ABSTRACT
Insulin-like growth factor receptor (IGF-1R) has been studied as an oncologic target in soft tissue sarcoma (STS), but its role in sarcoma biology is unclear. Anti-IGF-1R antibody cixutumumab demonstrated acceptable toxicity but limited activity as a single agent in STS. We carried out a dose-escalation study of cixutumumab with doxorubicin to evaluate safety and dosing of the combination. Eligible patients with advanced STS were treated with cixutumumab intravenously on days 1/8/15 at one of three dose levels (A: 1 mg/kg, B: 3 mg/kg, C: 6 mg/kg) with doxorubicin at 75 mg/m(2) as a 48 h infusion on day 1 of a 21 day cycle. After six cycles of the combination, patients could receive cixutumumab alone. The Time-to-Event Continual Reassessment Method was used to estimate the probability of dose-limiting toxicity (DLT) and to assign patients to the dose with an estimated probability of DLT≤20%. Between September 2008 and January 2012, 30 patients with advanced STS received a median of six cycles of therapy (range <1-22). Two DLTs were observed, grade 3 mucositis (dose level B) and grade 4 hyperglycemia (dose level C). Grade 2 and 3 reduced left ventricular ejection fraction was seen in three and two patients, respectively. Five partial responses were observed, and estimated progression-free survival was 5.3 months (95% confidence interval 3.0-6.3) in 26 response-assessable patients. Immunohistochemical staining of 11 available tumor samples for IGF-1R and phospho-IGF-1R was not significantly different among responders and non-responders, and serum analysis of select single-nucleotide polymorphisms did not predict for cardiotoxicity. The maximum tolerated dose was doxorubicin 75 mg/m(2) on day 1 and cixitumumab 6 mg/kg on days 1/8/15 of a 21 day cycle. Cardiac toxicity was observed and should be monitored in subsequent studies, which should be considered in STS only if a predictive biomarker of benefit to anti-IGF-1R therapy is identified. ClinicalTrials.gov:NCT00720174.
Sigma-Aldrich
Anti-IGF-IR Antibody, clone JBW902, clone JBW902, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-IR/IGF1R (Tyr1158/Tyr1162/Tyr1163) Antibody, Upstate®, from rabbit