MilliporeSigma
  • Disruption of glial cell development by Zika virus contributes to severe microcephalic newborn mice.
Disruption of glial cell development by Zika virus contributes to severe microcephalic newborn mice.
Cell discovery (2018-08-08)
Cui Li, Qin Wang, Yisheng Jiang, Qing Ye, Dan Xu, Fei Gao, Jesse W Xu, Ruoke Wang, Xingliang Zhu, Lei Shi, Lei Yu, Fuchun Zhang, Weixiang Guo, Linqi Zhang, Cheng-Feng Qin, Zhiheng Xu
ABSTRACT
The causal link between Zika virus (ZIKV) infection and microcephaly has raised alarm worldwide. Microglial hyperplasia, reactive gliosis, and myelination delay have been reported in ZIKV-infected microcephalic fetuses. However, whether and how ZIKV infection affects glial cell development remain unclear. Here we show that ZIKV infection of embryos at the later stage of development causes severe microcephaly after birth. ZIKV infects the glial progenitors during brain development. Specifically, ZIKV infection disturbs the proliferation and differentiation of the oligodendrocyte progenitor cells and leads to the abolishment of oligodendrocyte development. More importantly, a single intraperitoneal injection of pregnant mice with a human monoclonal neutralizing antibody provides full protection against ZIKV infection and its associated damages in the developing fetuses. Our results not only provide more insights into the pathogenesis of ZIKV infection, but also present a new model for the preclinical test of prophylactic and therapeutic agents against ZIKV infection.
Sigma-Aldrich
Anti-Olig1 Antibody, ascites fluid, Chemicon®
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit