MilliporeSigma
Detection of human herpesvirus 6 by reverse transcription-PCR.
Journal of clinical microbiology (1999-10-19)
R A Norton, M T Caserta, C B Hall, K Schnabel, P Hocknell, S Dewhurst
ABSTRACT
The role of human herpesvirus 6 (HHV-6) in disease beyond primary infection remains unclear. We have developed and validated a new reverse transcription-PCR (RT-PCR) assay for HHV-6 that can determine the presence of HHV-6 in clinical specimens and differentiate between latent and replicating virus. Peripheral blood mononuclear cells from 109 children were evaluated for HHV-6 by RT-PCR, DNA PCR, and viral culture. Of these samples, 106 were suitable for analysis. A total of 20 samples were positive for HHV-6 by culture and DNA PCR, of which 19 were positive by RT-PCR (sensitivity, 95%). All 28 samples from children that were negative by viral culture, but positive by DNA PCR, were negative for viral transcripts by our RT-PCR assay. One positive RT-PCR result was observed in 56 samples that were negative by tissue culture and DNA PCR. This indicates a low rate of false-positive results (1.2%) and a specificity of 98.8%. This RT-PCR assay can reliably differentiate between latent and actively replicating HHV-6 and should allow insight into the pathogenesis of this ubiquitous virus.
Sigma-Aldrich
Anti-Herpes Virus 6 Antibody, 101kDa protein, clone C3108-103, clone C3108-103, Chemicon®, from mouse