MilliporeSigma
  • Light-activated kinases enable temporal dissection of signaling networks in living cells.
Light-activated kinases enable temporal dissection of signaling networks in living cells.
Journal of the American Chemical Society (2011-01-29)
Arnaud Gautier, Alexander Deiters, Jason W Chin
ABSTRACT
We report a general strategy for creating protein kinases in mammalian cells that are poised for very rapid activation by light. By photoactivating a caged version of MEK1, we demonstrate the specific, rapid, and receptor independent activation of an artificial subnetwork within the Raf/MEK/ERK pathway. Time-lapse microscopy allowed us to precisely characterize the kinetics of elementary steps in the signaling cascade and provided insight into adaptive feedback and rate-determining processes in the pathway.
Sigma-Aldrich
Methyl-o-nitropiperonyllysine, ≥95%
Pricing and availability is not currently available.