MilliporeSigma
  • Role for the splicing factor TCERG1 in Cajal body integrity and snRNP assembly.
Role for the splicing factor TCERG1 in Cajal body integrity and snRNP assembly.
Journal of cell science (2019-10-23)
Cristina Moreno-Castro, Silvia Prieto-Sánchez, Noemí Sánchez-Hernández, Cristina Hernández-Munain, Carlos Suñé
ABSTRACT
Cajal bodies are nuclear organelles involved in the nuclear phase of small nuclear ribonucleoprotein (snRNP) biogenesis. In this study, we identified the splicing factor TCERG1 as a coilin-associated factor that is essential for Cajal body integrity. Knockdown of TCERG1 disrupts the localization of the components of Cajal bodies, including coilin and NOLC1, with coilin being dispersed in the nucleoplasm into numerous small foci, without affecting speckles, gems or the histone locus body. Furthermore, the depletion of TCERG1 affects the recruitment of Sm proteins to uridine-rich small nuclear RNAs (snRNAs) to form the mature core snRNP. Taken together, the results of this study suggest that TCERG1 plays an important role in Cajal body formation and snRNP biogenesis.
Sigma-Aldrich
Anti-2,2,7-Trimethylguanosine Antibody, clone K121, clone K121, from mouse
Sigma-Aldrich
Monoclonal Anti-Splicing Factor SC-35 antibody produced in mouse, clone SC-35, ascites fluid