MilliporeSigma
  • Band engineering at the interface of all-inorganic CsPbI2Br solar cells.
Band engineering at the interface of all-inorganic CsPbI2Br solar cells.
Nanoscale (2019-07-26)
Jing Zhuang, Yuanzhi Wei, Yigang Luan, Ningli Chen, Peng Mao, Shaokui Cao, Jizheng Wang
ABSTRACT
An all-inorganic CsPbI2Br perovskite with excellent phase stability and thermal stability has been considered to be a promising candidate for photovoltaic application. However, low efficiency and high moisture sensitivity hinder its advancement. In this work, we exploit 4-bromobenzylamine hydriodate post-treatment on CsPbI2Br thin films to assist the extraction of holes and to block the flow of electrons to the hole transport layer through band engineering at the CsPbI2Br bulk/surface. We found through depth profile analysis that a small amount of BrBeAI permeates into the CsPbI2Br bulk and mainly locates at the CsPbI2Br grain boundaries. This treatment leads to an improved short-circuit current of CsPbI2Br solar cells and an enhanced efficiency from 13.10% to 14.63%. In addition, the incorporation of the hydrophobic organic component into perovskite films effectively enhances the moisture resistance. This result proves that utilizing organic ammonium salt to improve the performance of the device through band alignment is an effective strategy for all-inorganic perovskite solar cell optimization.