MilliporeSigma
  • Asymmetric Synthesis of Adjacent Tri- and Tetrasubstituted Carbon Stereocenters: Organocatalytic Aldol Reaction of an Hydantoin Surrogate with Azaarene 2-Carbaldehydes.
Asymmetric Synthesis of Adjacent Tri- and Tetrasubstituted Carbon Stereocenters: Organocatalytic Aldol Reaction of an Hydantoin Surrogate with Azaarene 2-Carbaldehydes.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-07-19)
June Izquierdo, Noémie Demurget, Aitor Landa, Tore Brinck, Jose M Mercero, Peter Dinér, Mikel Oiarbide, Claudio Palomo
ABSTRACT
A bifunctional amine/squaramide catalyst promoted direct aldol addition of an hydantoin surrogate to pyridine 2-carbaldehyde N-oxides to afford adducts bearing two vicinal tertiary/quaternary carbons in high diastereo- and enantioselectivity (d.r. up to >20:1; ee up to 98 %) is reported. Acid hydrolysis of adducts followed by reduction of the N-oxide group yields enantiopure carbinol-tethered quaternary hydantoin-azaarene conjugates with densely functionalized skeletons. DFT studies of the potential energy surface (B3LYP/6-31+G(d)+CPCM (dichloromethane)) of the reaction correlate the activity of different catalysts and support an intramolecular hydrogen-bond-assisted activation of the squaramide moiety in the transition state of the catalytic reaction.
Sigma-Aldrich
Hydantoin, 98%