MilliporeSigma
  • Adenosine-stimulated astroglial swelling in cat cerebral cortex in vivo with total inhibition by a non-diuretic acylaryloxyacid derivative.
Adenosine-stimulated astroglial swelling in cat cerebral cortex in vivo with total inhibition by a non-diuretic acylaryloxyacid derivative.
Journal of neurosurgery (1981-09-01)
R S Bourke, J B Waldman, H K Kimelberg, K D Barron, B D San Filippo, A J Popp, L R Nelson
ABSTRACT
The intact cerebral cortices of cats were exposed in vivo under normothermic conditions and superfused with isotonic artificial cerebrospinal fluid containing added 0.125 mM adenosine. This resulted in chloridecation-rich cerebrocortical swelling which was shown by electron microscopy to be associated with an expanded astroglial compartment. The addition of DCPIB, a non-diuretic acylaryloxyacid analogue of ethacrynic acid and an inhibitor of coupled chloride-cation transport in cerebral cortex in vitro, totally blocked astroglial swelling and the concomitant increases in tissue ion contents. These studies support our previous experiments on the mechanism of formation of astroglial swelling. The pathological consequences of astroglial swelling and the clinical applications of these findings are discussed.
Sigma-Aldrich
DCPIB, ≥98% (HPLC)