MilliporeSigma
  • Regulation of p53 stability and function by the deubiquitinating enzyme USP42.
Regulation of p53 stability and function by the deubiquitinating enzyme USP42.
The EMBO journal (2011-11-17)
Andreas K Hock, Arnaud M Vigneron, Stephanie Carter, Robert L Ludwig, Karen H Vousden
ABSTRACT
The p53 tumour suppressor protein is a transcription factor that prevents oncogenic progression by activating the expression of apoptosis and cell-cycle arrest genes in stressed cells. The stability of p53 is tightly regulated by ubiquitin-dependent degradation, driven mainly by the ubiquitin ligase MDM2. In this study, we have identified USP42 as a DUB that interacts with and deubiquitinates p53. USP42 forms a direct complex with p53 and controls level of ubiquitination during the early phase of the response to a range of stress signals. Although we do not find a clear role for USP42 in controlling either the basal or fully activated levels of p53, the function of USP42 is required to allow the rapid activation of p53-dependent transcription and a p53-dependent cell-cycle arrest in response to stress. These functions of USP42 are likely to contribute to the repair and recovery of cells from mild or transient damage.