MilliporeSigma
  • Demographic responses of Cladocerans (Cladocera) in relation to different concentrations of humic substances.
Demographic responses of Cladocerans (Cladocera) in relation to different concentrations of humic substances.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2019-08-09)
Jose Luis Gama Flores, Maria Elena Huidobro Salas, S S S Sarma, S Nandini
ABSTRACT
Cladocerans are constantly exposed to humic substances in nature, yet the effects of these substances on their survival and reproduction are not well known. Here, the effects of humic substances (20 and 40 mg L-1) (HS) on the life history variables of three common cladocerans, Ceriodaphnia dubia, Moina macrocopa, and Daphnia pulex were evaluated. The results showed that the effect of humic substances on the tested cladocerans is species-specific, affecting either survival, reproduction or both. For M. macrocopa, exposed to HS at a concentration of 40 mg L-1, the average lifespan and the life expectancy at birth were significantly reduced as compared to controls, but for C. dubia and D. pulex these parameters were increased. Gross reproductive rate was unaffected by the HS level for both D. pulex and M. macrocopa, but it was significantly higher for C. dubia. When compared to the corresponding controls, for HS-exposed cladocerans, the rate of population increase was significantly reduced in case of D. pulex while it was stimulated for both C. dubia and M. macrocopa. It appears that humic substances had a slightly stronger influence on survivorship than on reproduction of the tested cladocerans.