MilliporeSigma
The role of thrombin in the pathogenesis of diabetic neuropathy.
PloS one (2019-07-06)
Efrat Shavit-Stein, Ramona Aronovich, Constantin Sylantiev, Shany Guly Gofrit, Joab Chapman, Amir Dori
ABSTRACT
Diabetic neuropathy is common and disabling despite glycemic control. Novel neuroprotective approaches are needed. Thrombin and hypercoagulability are associated with diabetes and nerve conduction dysfunction. Our aim was to study the role of thrombin in diabetic neuropathy. We measured thrombin activity by a biochemical assay in streptozotocin (STZ)-induced diabetic neuropathy in male Sprague-Dawley rats. Neuropathy severity was assessed by thermal latency and nerve conduction measures. Thermal latencies were longer in diabetic rats, and improved with the non-specific serine-protease inhibitor Tosyl-L-lysine-chloromethyl ketone (TLCK) treatment (p<0.01). The tail nerve of diabetic rats showed slow conduction velocity (p˂0.01), and interestingly, increased thrombin activity was noted in the sciatic nerve (p˂0.001). Sciatic nodes of Ranvier and the thrombin receptor, protease activated receptor 1 (PAR1) reactivity showed abnormal morphology in diabetic animals by immunofluorescence staining (p<0.0001). Treatment of diabetic animals with either the specific thrombin inhibitor, N-alpha 2 naphtalenesulfonylglycyl alpha-4 amidino-phenylalaninepiperidide (NAPAP) or TLCK preserved normal conduction velocity, (p˂0.01 and p = 0.01 respectively), and prevented disruption of morphology (p˂0.05 and p˂0.03). The results establish for the first time an association between diabetic neuropathy and excessive activation of the thrombin pathway. Treatment of diabetic animals with thrombin inhibitors ameliorates both biochemical, structural and electrophysiological deficits. The thrombin pathway inhibition may be a novel neuroprotective therapeutic target in the diabetic neuropathy pathology.
Sigma-Aldrich
N-p-Tosyl-Gly-Pro-Arg 7-amido-4-methylcoumarin hydrochloride, protease substrate