MilliporeSigma
  • Is FAS/Fas ligand system involved in equine corpus luteum functional regression?
Is FAS/Fas ligand system involved in equine corpus luteum functional regression?
Biology of reproduction (2010-08-20)
Antonio M Galvao, David W Ramilo, Dariusz J Skarzynski, Karolina Lukasik, Angela Tramontano, Antonio Mollo, Luisa M Mateus, Graca Maria L Ferreira-Dias
ABSTRACT
Proapoptotic factor Fas ligand (FASL) and its cell surface receptor FAS are tumor necrosis factor superfamily members that trigger apoptosis in different cell types. However, their influence on luteal steroidogenesis is not clearly understood. The aim of the present work was to determine (i) the presence of the cytokine FASL and its receptor FAS in the mare's corpus luteum (CL) throughout the luteal phase, as well as (ii) the influence of FASL alone, or together with the cytokines tumor necrosis factor alpha (TNF) and interferon gamma (IFNG), on equine luteal cell production of luteotrophic and luteolytic factors, cell viability, and apoptosis. FASL and FAS protein expression and mRNA transcription were evaluated in different luteal stages of the equine CL by Western blotting and real-time PCR assays, respectively. Protein expression and FASL mRNA transcription increased in the late CL. Also, FAS and FASL proteins were present in large steroidogenic and endothelial CL cells throughout the luteal phase, as demonstrated by immunohistochemistry. Equine luteal cells isolated from midluteal phase CL were stimulated without (control) or with exogenous cytokines: FASL (10 ng/ml); TNF+IFNG (10 ng/ml each; positive control) or FASL+TNF+IFNG (10 ng/ml each). FASL clearly inhibited in vitro progesterone and prostaglandin E(2) (PGE(2)) production by equine luteal cells but increased prostaglandin F(2alpha) (PGF(2alpha)). Furthermore, FASL effect on equine luteal cell viability depended on the presence of cytokines TNF and IFNG. In conclusion, this study shows the presence of FASL and FAS in the equine CL and suggests their importance in functional luteolysis.
Sigma-Aldrich
Fas Ligand human, >95% (SDS-PAGE), recombinant, expressed in CHO cells, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.