MilliporeSigma
Photocrosslinkable polysaccharides for in situ hydrogel formation.
Journal of biomedical materials research (2000-11-15)
K A Smeds, A Pfister-Serres, D Miki, K Dastgheib, M Inoue, D L Hatchell, M W Grinstaff
ABSTRACT
In situ photopolymerization is an exciting new technique for tissue engineering. Two photocrosslinkable polysaccharides composed of alginate and hyaluronan are described that upon photolysis form soft, flexible, and viscoelastic hydrogels. The degree of methacrylate modification and thus covalent affects mechanical properties such as swelling, compression, and creep compliance. Significant swelling is observed in aqueous solution; these hydrogels can swell up to 14 times their dry weight. Both hydrogels exhibit low phase angles and (G*) values indicative of viscoelastic materials. The hyaluronan based hydrogel is stronger and more resilient than the corresponding alginate gel. SEM and AFM studies on both hydrogels show smooth and uniform surfaces at the macroscopic level with salient features observed only on the nanometer scale. Rapid polymerization by an optical trigger allows for controlled in situ photopolymerization in a minimally invasive manner, indicating that these hydrogels are relevant for biomedical applications such as sealing wounds and reconstructing soft tissues.
Sigma-Aldrich
Methacrylated Alginate, Medium Viscosity, Low endotoxin