MilliporeSigma
  • NHE3 activity is dependent on direct phosphoinositide binding at the N terminus of its intracellular cytosolic region.
NHE3 activity is dependent on direct phosphoinositide binding at the N terminus of its intracellular cytosolic region.
The Journal of biological chemistry (2010-08-26)
Sachin Mohan, Chung Ming Tse, Sandra B Gabelli, Rafiquel Sarker, Boyoung Cha, Kamau Fahie, Mythili Nadella, Nicholas C Zachos, Becky Tu-Sekine, Daniel Raben, L Mario Amzel, Mark Donowitz
ABSTRACT
The small intestinal BB Na(+)/H(+) antiporter NHE3 accounts for the majority of intestinal sodium and water absorption. It is highly regulated with both postprandial inhibition and stimulation sequentially occurring. Phosphatidylinositide 4,5-bisphosphate (PI(4,5)P(2)) and phosphatidylinositide 3,4,5-trisphosphate (PI(3,4,5)P(3)) binding is involved with regulation of multiple transporters. We tested the hypothesis that phosphoinositides bind NHE3 under basal conditions and are necessary for its acute regulation. His(6) proteins were made from the NHE3 C-terminal region divided into four parts as follows: F1 (amino acids 475-589), F2 (amino acids 590-667), F3 (amino acids 668-747), and F4 (amino acids 748-832) and purified by a nickel column. Mutations were made in the F1 region of NHE3 and cloned in pet30a and pcDNA3.1 vectors. PI(4,5)P(2) and PI(3,4,5)P(3) bound only to the NHE3 F1 fusion protein (amino acids 475-589) on liposomal pulldown assays. Mutations were made in the putative lipid binding region of the F1 domain and studied for alterations in lipid binding and Na(+)/H(+) exchange as follows: Y501A/R503A/K505A; F509A/R511A/R512A; R511L/R512L; R520/FR527F; and R551L/R552L. Our results indicate the following. 1) The F1 domain of the NHE3 C terminus has phosphoinositide binding regions. 2) Mutations of these regions alter PI(4,5)P(2) and PI(3,4,5)P(3) binding and basal NHE3 activity. 3) The magnitude of serum stimulation of NHE3 correlates with PI(4,5)P(2) and PI(3,4,5)P(3) binding of NHE3. 4) Wortmannin inhibition of PI3K did not correlate with PI(4,5)P(2) or PI(3,4,5)P(3) binding of NHE3. Two functionally distinct phosphoinositide binding regions (Tyr(501)-Arg(512) and Arg(520)-Arg(552)) are present in the NHE3 F1 domain; both regions are important for serum stimulation, but they display differences in phosphoinositide binding, and the latter but not the former alters NHE3 surface expression.
Product Description
Millipore
Anti-VSV-Glycoprotein−Agarose antibody, Mouse monoclonal, clone P5D4, purified from hybridoma cell culture, PBS suspension