MilliporeSigma
  • The application of a photon-counting camera in sensitive, bioluminescence-enhanced detection systems for nucleic acid hybridization. Ultrasensitive detection systems for protein blotting and DNA hybridization, III.
The application of a photon-counting camera in sensitive, bioluminescence-enhanced detection systems for nucleic acid hybridization. Ultrasensitive detection systems for protein blotting and DNA hybridization, III.
Journal of clinical chemistry and clinical biochemistry. Zeitschrift fur klinische Chemie und klinische Biochemie (1989-06-01)
R Hauber, R Geiger
ABSTRACT
A relatively simple, bioluminescence-enhanced detection system for nucleic acid hybridization, using alkaline phosphatase as a label, was described recently (Hauber, R. & Geiger, R. (1988) Nucl. Acid Res. 16, 1213). The principle of detection is as follows: Alkaline phosphatase releases D-luciferin (Photinus pyralis) from D-luciferin-O-phosphate. Liberated D-luciferin reacts with luciferase, ATP and oxygen with light emission. Light produced is measured with a very sensitive photon counting camera system (Argus-100), allowing the visualization and localization of the specifically bound alkaline phosphatase on nitrocellulose sheets. Under non-optimized conditions the limit of detection is at present about 30 pg of pBR322. A sulphonylated nucleotide probe was used for hybridization.