MilliporeSigma
  • Discovery of quinazolines as a novel structural class of potent inhibitors of NF-kappa B activation.
Discovery of quinazolines as a novel structural class of potent inhibitors of NF-kappa B activation.
Bioorganic & medicinal chemistry (2003-01-09)
Masanori Tobe, Yoshiaki Isobe, Hideyuki Tomizawa, Takahiro Nagasaki, Hirotada Takahashi, Tominaga Fukazawa, Hideya Hayashi
ABSTRACT
We disclose here a new structural class of low-molecular-weight inhibitors of NF-kappa B activation that were designed and synthesized by starting from quinazoline derivative 6a. Structure-activity relationship (SAR) studies based on 6a elucidated the structural requirements essential for the inhibitory activity toward NF-kappa B transcriptional activation, and led to the identification of the 6-amino-4-phenethylaminoquinazoline skeleton as the basic framework. In this series of compounds, 11q, containing the 4-phenoxyphenethyl moiety at the C(4)-position, showed strong inhibitory effects on both NF-kappa B transcriptional activation and TNF-alpha production. Furthermore, 11q exhibited an anti-inflammatory effect on carrageenin-induced paw edema in rats.
Sigma-Aldrich
NF-κB Activation Inhibitor, InSolution, ≥98%