MilliporeSigma
  • Small molecule inhibition of protein depalmitoylation as a new approach towards downregulation of oncogenic Ras signalling.
Small molecule inhibition of protein depalmitoylation as a new approach towards downregulation of oncogenic Ras signalling.
Bioorganic & medicinal chemistry (2010-12-07)
Frank J Dekker, Christian Hedberg
ABSTRACT
The H- and N-Ras GTPases are prominent examples of proteins, whose localizations and signalling capacities are regulated by reversible palmitoylations and depalmitoylations. Recently, the novel small molecule inhibitor palmostatin B has been described to inhibit Ras depalmitoylation and to revert the phenotype of oncogenic HRasG12V transformed cells. This demonstrates that palmostatin B is a tool to investigate the biochemical effects of the inhibition of cellular Ras depalmitoylation on Ras signalling, which is relevant for oncology. Furthermore, it is to be expected that many proteins, of which the signalling capacities depend on reversible palmitoylation, will be discovered in the near future. This stresses the urgent need for further development of small molecule inhibitors of palmitoylation and depalmitoylation in order to study their functions in cellular signalling.
Sigma-Aldrich
Palmostatin B, InSolution, ≥95%, 50 mM in DMSO, APT1 inhibitor
Pricing and availability is not currently available.