MilliporeSigma
  • The biflavonoid isoginkgetin is a general inhibitor of Pre-mRNA splicing.
The biflavonoid isoginkgetin is a general inhibitor of Pre-mRNA splicing.
The Journal of biological chemistry (2008-10-02)
Kristine O'Brien, Arianne J Matlin, April M Lowell, Melissa J Moore
ABSTRACT
Membrane-permeable compounds that reversibly inhibit a particular step in gene expression are highly useful tools for cell biological and biochemical/structural studies. In comparison with other gene expression steps where multiple small molecule effectors are available, very few compounds have been described that act as general inhibitors of pre-mRNA splicing. Here we report construction and validation of a set of mammalian cell lines suitable for the identification of small molecule inhibitors of pre-mRNA splicing. Using these cell lines, we identified the natural product isoginkgetin as a general inhibitor of both the major and minor spliceosomes. Isoginkgetin inhibits splicing both in vivo and in vitro at similar micromolar concentrations. It appears to do so by preventing stable recruitment of the U4/U5/U6 tri-small nuclear ribonucleoprotein, resulting in accumulation of the prespliceosomal A complex. Like two other recently reported general pre-mRNA splicing inhibitors, isoginkgetin has been previously described as an anti-tumor agent. Our results suggest that splicing inhibition is the mechanistic basis of the anti-tumor activity of isoginkgetin. Thus, pre-mRNA splicing inhibitors may represent a novel avenue for development of new anti-cancer agents.
Sigma-Aldrich
Pre-mRNA Splicing Inhibitor, Isoginkgetin, The Pre-mRNA Splicing Inhibitor, Isoginkgetin, also referenced under CAS 548-19-6, blocks the spliceosome-meidated splicing process.
Pricing and availability is not currently available.