MilliporeSigma
  • bcl-x, a bcl-2-related gene that functions as a dominant regulator of apoptotic cell death.
bcl-x, a bcl-2-related gene that functions as a dominant regulator of apoptotic cell death.
Cell (1993-08-27)
L H Boise, M González-García, C E Postema, L Ding, T Lindsten, L A Turka, X Mao, G Nuñez, C B Thompson
ABSTRACT
We report the isolation of bcl-x, a bcl-2-related gene that can function as a bcl-2-independent regulator of programmed cell death (apoptosis). Alternative splicing results in two distinct bcl-x mRNAs. The protein product of the larger mRNA, bcl-xL, is similar in size and predicted structure to Bcl-2. When stably transfected into an IL-3-dependent cell line, bcl-xL inhibits cell death upon growth factor withdrawal at least as well as bcl-2. Surprisingly, the second mRNA species, bcl-xS, encodes a protein that inhibits the ability of bcl-2 to enhance the survival of growth factor-deprived cells. In vivo, bcl-xS mRNA is expressed at high levels in cells that undergo a high rate of turnover, such as developing lymphocytes. In contrast, bcl-xL is found in tissues containing long-lived postmitotic cells, such as adult brain. Together these data suggest that bcl-x plays an important role in both positive and negative regulation of programmed cell death.