MilliporeSigma
  • Ibuprofen protects dopaminergic neurons against glutamate toxicity in vitro.
Ibuprofen protects dopaminergic neurons against glutamate toxicity in vitro.
Neuroscience letters (2000-08-29)
D Casper, U Yaparpalvi, N Rempel, P Werner
ABSTRACT
Non-steroidal anti-inflammatory drugs (NSAIDs) reduce the risk of Alzheimer's disease, although the underlying mechanisms are unknown. Glutamate excitotoxicity has been implicated in Alzheimer's disease, Parkinson's disease, and others. We examined the effects of aspirin, acetaminophen, and ibuprofen on cultured primary rat embryonic neurons from mesencephalon, the area primarily affected in Parkinson's disease. We evaluated whether these drugs protect dopaminergic neurons against excitotoxicity. All three NSAIDs significantly attenuated the decrease in dopamine uptake caused by glutamate, indicating preservation of neuronal integrity. One hundred micro-moles ibuprofen protected both dopaminergic neurons and neurons overall against glutamate toxicity. In addition, ibuprofen alone increased the relative number of dopaminergic neurons by 47%. Thus, NSAIDs protected neurons against glutamate excitotoxicity in vitro, and deserve further consideration as neuroprotective agents in Parkinson's disease.
Sigma-Aldrich
(±)-Ibuprofen, A nonsteroidal anti-inflammatory drug (NSAID) that acts as a reversible and competitive inhibitor of cyclooxygenase 1 (COX-1) (IC₅₀ = 4.85 µM).