MilliporeSigma
  • Identification of distinct endoglycosidase (endo) activities in Flavobacterium meningosepticum: endo F1, endo F2, and endo F3. Endo F1 and endo H hydrolyze only high mannose and hybrid glycans.
Identification of distinct endoglycosidase (endo) activities in Flavobacterium meningosepticum: endo F1, endo F2, and endo F3. Endo F1 and endo H hydrolyze only high mannose and hybrid glycans.
The Journal of biological chemistry (1991-01-25)
R B Trimble, A L Tarentino
ABSTRACT
Flavobacterium meningosepticum endo-beta-acetyl-glucosaminidase F preparations have been resolved by hydrophobic interaction chromatography on TSK-butyl resin into at least three activities designated endo F1, endo F2 and endo F3 each with a unique substrate specificity. The 32-kDa endo F1 protein is the principle component representing in excess of 95% of most earlier and currently available commercial endoglycosidase preparations, the remainder being a mixture of five proteins from 32 to 43 kDa. Substrate specificity studies reveal endo F1 and endo H from Streptomyces plicatus to have nearly identical capacities to hydrolyze high-mannose oligosaccharides with a minimum Man1 alpha 3Man1 alpha 6Man1 beta 4GlcNAc1 beta 4GlcNAc structure. Although endo H will hydrolyze fucose-containing hybrid oligosaccharides at rates approaching comparable high-mannose forms, core-linked fucose reduces the hydrolysis rate of endo F1 by over 50-fold relative to high-mannose structures. Neither homogeneous endo F1 nor endo H hydrolyze complex multi-antennary glycans. The biantennary cleaving activity previously reported for endo F preparations (Tarentino, A. L., Gómez, C. M., and Plummer, T. H., Jr. (1985) Biochemistry 24, 4665-4671) is a characteristic of the contaminating endo F2 activity.
Sigma-Aldrich
Endoglycosidase F2, Elizabethkingia meningosepticum, Recombinant, E. coli, Endoglycosidase F2, Elizabethkingia meningosepticum, Recombinant, E. coli cleaves asparagine-linked or free oligomannose and biantennary complex oligosaccharides. It is not active above pH 6.0.
Pricing and availability is not currently available.
Sigma-Aldrich
Endoglycosidase F1, Elizabethkingia meningosepticum, Recombinant, E. coli, Endoglycosidase F1, Elizabethkingia meningosepticum, Recombinant, E. coli cleaves asparagine-linked or free oligomannose and hybrid. Suitable for deglycosylation of native proteins.
Pricing and availability is not currently available.
Sigma-Aldrich
Endoglycosidase F3, Elizabethkingia meningosepticum, Recombinant, E. coli, Endoglycosidase F3, Elizabethkingia meningosepticum, Recombinant, E. coli, cleaves asparagine-linked or free biantennary and triantennary complex, and Man3GlcNAc oligosaccharides from glycoproteins.
Pricing and availability is not currently available.