MilliporeSigma
  • Boosting subdominant neutralizing antibody responses with a computationally designed epitope-focused immunogen.
Boosting subdominant neutralizing antibody responses with a computationally designed epitope-focused immunogen.
PLoS biology (2019-02-23)
Fabian Sesterhenn, Marie Galloux, Sabrina S Vollers, Lucia Csepregi, Che Yang, Delphyne Descamps, Jaume Bonet, Simon Friedensohn, Pablo Gainza, Patricia Corthésy, Man Chen, Stéphane Rosset, Marie-Anne Rameix-Welti, Jean-François Éléouët, Sai T Reddy, Barney S Graham, Sabine Riffault, Bruno E Correia
ABSTRACT
Throughout the last several decades, vaccination has been key to prevent and eradicate infectious diseases. However, many pathogens (e.g., respiratory syncytial virus [RSV], influenza, dengue, and others) have resisted vaccine development efforts, largely because of the failure to induce potent antibody responses targeting conserved epitopes. Deep profiling of human B cells often reveals potent neutralizing antibodies that emerge from natural infection, but these specificities are generally subdominant (i.e., are present in low titers). A major challenge for next-generation vaccines is to overcome established immunodominance hierarchies and focus antibody responses on crucial neutralization epitopes. Here, we show that a computationally designed epitope-focused immunogen presenting a single RSV neutralization epitope elicits superior epitope-specific responses compared to the viral fusion protein. In addition, the epitope-focused immunogen efficiently boosts antibodies targeting the palivizumab epitope, resulting in enhanced neutralization. Overall, we show that epitope-focused immunogens can boost subdominant neutralizing antibody responses in vivo and reshape established antibody hierarchies.