MilliporeSigma
  • Generation of induced pluripotent stem cell lines from two Neuroblastoma patients carrying a germline ALK R1275Q mutation.
Generation of induced pluripotent stem cell lines from two Neuroblastoma patients carrying a germline ALK R1275Q mutation.
Stem cell research (2019-01-04)
Ana Marin Navarro, Kelly Day, Per Kogner, Margareta Wilhelm, Anna Falk
ABSTRACT
Neuroblastoma (NB) is an embryonic tumor of the peripheral nervous system and one of the most common solid cancers in infants. Mutations in the Anaplastic lymphoma tyrosine kinase (ALK) gene are common in NB. To study the contribution of ALK mutations in NB initiation and progression, we reprogrammed fibroblasts from two related NB patients carrying germline mutations in ALK (R1275Q) using non-integrating Sendai virus. The iPS cells are grown in a feeder- and xeno-free conditions, have normal karyotype, retain the ALK R1275Q mutation, have been characterized by expression of pluripotency markers and differentiation to all three germ layers.