MilliporeSigma
  • A trypsin inhibitor-like protein secreted by Cotesia vestalis teratocytes inhibits hemolymph prophenoloxidase activation of Plutella xylostella.
A trypsin inhibitor-like protein secreted by Cotesia vestalis teratocytes inhibits hemolymph prophenoloxidase activation of Plutella xylostella.
Journal of insect physiology (2019-04-27)
Qi-Juan Gu, Shi-Min Zhou, Yue-Nan Zhou, Jian-Hua Huang, Min Shi, Xue-Xin Chen
ABSTRACT
To establish successful infections, endoparasitoid wasps must develop strategies to evade immune responses of the host. Here, we identified and characterized a teratocytes-expressed gene encoding a trypsin inhibitor-like protein containing a cysteine-rich domain from Cotesia vestalis, CvT-TIL. CvT-TIL had a high expression level during the later developmental stage of teratocytes and was secreted into host hemolymph. Further experiments showed CvT-TIL strongly suppressed the prophenoloxidase activation of host hemolymph in a dose-dependent manner by interacting with PxPAP3 of PO cascade. Our results not only provide evidence for an inhibition between CvT-TIL gene and the host's melanization activity, but also expand our knowledge about the mechanisms by which parasitoids regulate humoral immunity of the host.