MilliporeSigma
  • Temperature-modulated cell-separation column using temperature-responsive cationic copolymer hydrogel-modified silica beads.
Temperature-modulated cell-separation column using temperature-responsive cationic copolymer hydrogel-modified silica beads.
Colloids and surfaces. B, Biointerfaces (2019-03-16)
Kenichi Nagase, Daimu Inanaga, Daiju Ichikawa, Aya Mizutani Akimoto, Yutaka Hattori, Hideko Kanazawa
ABSTRACT
There is strong demand for cell separation methods that do not decrease cell activity or modify cell surfaces. Here, new temperature-modulated cell-separation columns not requiring cell-surface premodification are described. The columns were packed with temperature-responsive cationic polymer hydrogel-modified silica beads. Poly(N-isopropylacrylamide-co-n-butyl methacrylate-co-N,N-dimethylaminopropyl acrylamide) hydrogels with various cationic moieties were attached to silica-bead surfaces by radical polymerization using N,N'-methylenebisacrylamide as a crosslinking agent. The beads were packed into solid-phase extraction columns, and temperature-dependent cell elution from the columns was found using HL-60 and Jurkat cells. The retention HL-60 and Jurkat cells in columns containing cationic beads at 37 °C was 95.3% to 99.6% and 95.0% to 98.8%, respectively. By contrast, beads without cationic properties exhibited low cell retention (20.6% for HL-60 and 32.5% for Jurkat cells). The cells were mainly retained through both electrostatic and hydrophobic interactions. The retained HL-60 (4.9%) and Jurkat cells (40%) were eluted at 4 °C from the column with a low composition of cationic monomer (DMAPAAm, 1 mol% in copolymer), because the temperature-responsive hydrogels on the beads became hydrophilic, decreasing the hydrophobic interactions between the cells and the beads. A higher number of Jurkat cells than HL-60 cells were eluted because of differences in their electrostatic properties (Jurkat cells: -2.53 mV; HL-60 cells: -20.7 mV). The results indicated that cell retention by the hydrogel-coated beads packed in a solid phase extraction column could be modulated simply by changing the temperature.