- A New Bioactive Glass/Collagen Hybrid Composite for Applications in Dentistry.
A New Bioactive Glass/Collagen Hybrid Composite for Applications in Dentistry.
Bioactive glasses (BGs) are currently employed in a wide range of medical and dentistry applications by virtue of their bone-bonding ability. The incorporation of BGs into a collagen matrix may be used to combine the regenerative potential of these materials with the specific biological advantages of collagen. However, most of the collagen/BG composites reported in the literature are scaffolds and there is a lack of moldable putties or injectable systems. Here, granules of an innovative BG containing strontium and magnesium were mixed with collagen and PEG to obtain a putty (BGMS/C) suitable for dental applications. For the sake of comparison, granules of 45S5 Bioglass®, the gold standard among BGs, were used to prepare a 45S5/collagen putty. Both the composites were evaluated in vitro with respect to murine fibroblasts. The materials showed an excellent biocompatibility, making them interesting for possible applications in dentistry and reconstructive surgery. Moreover, BGMS/C seems to stimulate cell proliferation.