MilliporeSigma
  • Photo actuation of liquid crystalline elastomer nanocomposites incorporated with gold nanoparticles based on surface plasmon resonance.
Photo actuation of liquid crystalline elastomer nanocomposites incorporated with gold nanoparticles based on surface plasmon resonance.
Soft matter (2019-07-10)
Jiaojiao Xu, Shuang Chen, Wenlong Yang, Ban Qin, Xiuxiu Wang, Yuchang Wang, Maosheng Cao, Yachen Gao, Chensha Li, Yinmao Dong
ABSTRACT
In this work, according to the characteristic of surface plasmon resonance (SPR) of metallic nanoparticles, we investigated the photo actuation performance of a liquid crystalline elastomer (LCE) nanocomposite with incorporated gold nanoparticles (nano-gold/LCE nanocomposite). The nano-gold/LCE nanocomposites were fabricated by incorporating gold nanoparticles into a polysiloxane-based LCE matrix via a novel experimental protocol, and characterized by a well-developed SPR absorption band in the visible spectrum range. The nano-gold/LCE nanocomposites demonstrated strong actuation upon irradiation with a quasi-daylight source; the reason lay in that the SPR response of gold nanoparticles performed efficient energy conversion from light energy to thermal energy, and thus offered an activation pathway for the nematic-isotropic transition of the LCE matrix. The nano-gold/LCE nanocomposites underwent rapid maximum axial contraction up to about one third of the original length under light irradiation, and this photo-stimulated muscle-like actuation was fully reversible via the on-off switching of the light source. The photo actuation properties of nano-gold/LCE nanocomposites with varying irradiation intensities and gold nanoparticle content were also investigated. In addition, the nano-gold/LCE nanocomposites demonstrated superior optical nonlinear properties, and revealed potential for the application area of mode-locking for laser technology.