MilliporeSigma
  • Discovery of a Pathogenic Variant rs139379666 (p. P2974L) in ATM for Breast Cancer Risk in Chinese Populations.
Discovery of a Pathogenic Variant rs139379666 (p. P2974L) in ATM for Breast Cancer Risk in Chinese Populations.
Cancer epidemiology, biomarkers & prevention : a publication of the American Association for Cancer Research, cosponsored by the American Society of Preventive Oncology (2019-06-05)
Xingyi Guo, Weiqiang Lin, Mengqiu Bai, Hongzhi Li, Wanqing Wen, Chenjie Zeng, Zhishan Chen, Jing He, Jianghua Chen, Qiuyin Cai, Jirong Long, Wei-Hua Jia, Xiao-Ou Shu, Wei Zheng
ABSTRACT
Pathogenic variants in susceptibility genes lead to increased breast cancer risk. To identify coding variants associated with breast cancer risk, we conducted whole-exome sequencing in genomic DNA samples from 831 breast cancer cases and 839 controls of Chinese women. We also genotyped samples, including 4,580 breast cancer cases and 6,695 controls, using whole exome-chip arrays. We further performed a replication study using a Multi-Ethnic Global Array in samples from 1,793 breast cases and 2,059 controls. A single marker analysis was performed using the Fisher exact test. We identified a missense variant (rs139379666, P2974L; AF = 0.09% for breast cancer cases, but none for controls) in the ATM gene for breast cancer risk using combing data from 7,204 breast cancer cases and 9,593 controls (P = 1.7 × 10-5). To investigate the functionality of the variant, we first silenced ATM and then transfected the overexpression vectors of ATM containing the risk alleles (TT) or reference alleles (CC) of the variant in U2OS and breast cancer SK-BR3 cells, respectively. Our results showed that compared with the reference allele, the risk allele significantly disrupts the activity of homologous recombination-mediated double-strand breaks repair efficiency. Our results further showed that the risk allele may play a defected regulation role in the activity of the ATM structure. Our findings identified a novel mutation that disrupts ATM function, conferring to breast cancer risk. Functional investigation of genetic association findings is necessary to discover a pathogenic variant for breast cancer risk.