MilliporeSigma
  • Clear and transparent nanocrystals for infrared-responsive carrier transfer.
Clear and transparent nanocrystals for infrared-responsive carrier transfer.
Nature communications (2019-01-27)
Masanori Sakamoto, Tokuhisa Kawawaki, Masato Kimura, Taizo Yoshinaga, Junie Jhon M Vequizo, Hironori Matsunaga, Chandana Sampath Kumara Ranasinghe, Akira Yamakata, Hiroyuki Matsuzaki, Akihiro Furube, Toshiharu Teranishi
ABSTRACT
Infrared-light-induced carrier transfer is a key technology for 'invisible' optical devices for information communication systems and energy devices. However, clear and colourless photo-induced carrier transfer has not yet been demonstrated in the field of photochemistry, to the best of our knowledge. Here, we resolve this problem by employing short-wavelength-infrared (1400-4000 nm) localized surface plasmon resonance-induced electron injection from indium tin oxide nanocrystals to transparent metal oxides. The time-resolved infrared measurements visualize the dynamics of the carrier in this invisible system. Selective excitation of localized surface plasmon resonances causes hot electron injection with high efficiency (33%) and long-lived charge separation (~ 2-200 μs). We anticipate our study not only provides a breakthrough for plasmonic carrier transfer systems but may also stimulate the invention of state-of-the-art invisible optical devices.