MilliporeSigma
  • Desmoglein 2 mutation provokes skeletal muscle actin expression and accumulation at intercalated discs in murine hearts.
Desmoglein 2 mutation provokes skeletal muscle actin expression and accumulation at intercalated discs in murine hearts.
Journal of cell science (2019-01-20)
Sebastian Kant, Benjamin Freytag, Antonia Herzog, Anna Reich, Rudolf Merkel, Bernd Hoffmann, Claudia A Krusche, Rudolf E Leube
ABSTRACT
Arrhythmogenic cardiomyopathy (AC) is an incurable progressive disease that is linked to mutations in genes coding for components of desmosomal adhesions that are localized to the intercalated disc region, which electromechanically couples adjacent cardiomyocytes. To date, the underlying molecular dysfunctions are not well characterized. In two murine AC models, we find an upregulation of the skeletal muscle actin gene (Acta1), which is known to be a compensatory reaction to compromised heart function. Expression of this gene is elevated prior to visible morphological alterations and clinical symptoms, and persists throughout pathogenesis with an additional major rise during the chronic disease stage. We provide evidence that the increased Acta1 transcription is initiated through nuclear activation of the serum response transcription factor (SRF) by its transcriptional co-activator megakaryoblastic leukemia 1 protein (MKL1, also known as MRTFA). Our data further suggest that perturbed desmosomal adhesion causes Acta1 overexpression during the early stages of the disease, which is amplified by transforming growth factor β (TGFβ) release from fibrotic lesions and surrounding cardiomyocytes during later disease stages. These observations highlight a hitherto unknown molecular AC pathomechanism.
Sigma-Aldrich
Anti-ACTIN-α-1 antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution