MilliporeSigma
  • The Wave complex controls epidermal morphogenesis and proliferation by suppressing Wnt-Sox9 signaling.
The Wave complex controls epidermal morphogenesis and proliferation by suppressing Wnt-Sox9 signaling.
The Journal of cell biology (2019-03-15)
Jonathan Cohen, Shaul Raviv, Orit Adir, Krishnanand Padmanabhan, Arad Soffer, Chen Luxenburg
ABSTRACT
Development of the skin epidermis requires tight spatiotemporal control over the activity of several signaling pathways; however, the mechanisms that orchestrate these events remain poorly understood. Here, we identify a key role for the Wave complex proteins ABI1 and Wave2 in regulating signals that control epidermal shape and growth. In utero RNAi-mediated silencing of Abi1 or Wasf2 induced cellular hyperproliferation and defects in architecture of the interfollicular epidermis (IFE) and delayed hair follicle growth. Unexpectedly, SOX9, a hair follicle growth regulator, was aberrantly expressed throughout the IFE of the mutant embryos, and its forced overexpression mimicked the Wave complex loss-of-function phenotype. Moreover, Wnt signaling, which regulates SOX9+ cell specification, was up-regulated in Wave complex loss-of-function IFE. Importantly, we show that the Wave complex regulates filamentous actin content and that a decrease in actin levels is sufficient to elevate Wnt/β-catenin signaling. Our results identify a novel role for Wave complex- and actin-regulated signaling via Wnt and SOX9 in skin development.
Sigma-Aldrich
Anti-α-E-Catenin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
XAV939, ≥98% (HPLC)