MilliporeSigma
  • Androgen receptor is a negative regulator of contextual fear memory in male mice.
Androgen receptor is a negative regulator of contextual fear memory in male mice.
Hormones and behavior (2018-09-03)
Firyal Ramzan, Amber B Azam, D Ashley Monks, Iva B Zovkic
ABSTRACT
Although sex-hormones have a well-documented role in memory formation, most literature has focused on estrogens, whereas the role of androgens and their receptor (the androgen receptor; AR) in fear memory is relatively unexplored. To address this gap, we used a transgenic mouse model of AR overexpression (CMV-AR) to determine if AR regulates fear memory, and if this effect can be reversed either by the removal of circulating androgens via gonadectomy, or by antagonising AR activity with flutamide. We found that AR overexpression results in reduced freezing in response to foot shock, and that this difference is reversed with both gonadectomy and flutamide treatment. Differences between genotypes were reinstated by testosterone replacement in gonadectomized mice, suggesting that reduced fear memory in mutants results from AR activation by testosterone and is not secondary to group differences in circulating testosterone. Potential transcriptional mechanisms by which CMV-AR exerts its effects on fear memory were assessed by quantitating the expression of memory-related genes in area CA1 of the hippocampus. Several genes that are altered with AR inhibition and activation, including genes that encode for the histone variant H2A.Z, cholinergic receptors, glutamate receptors, and brain-derived neurotrophic factor. Overall, our findings suggest that AR is a negative regulator of fear memory and identify potential gene targets through which AR may mediate this effect.