MilliporeSigma
  • Thorium decorporation efficacy of rationally-selected biocompatible compounds with relevance to human application.
Thorium decorporation efficacy of rationally-selected biocompatible compounds with relevance to human application.
Journal of hazardous materials (2019-01-09)
Manjoor Ali, Biswajit Sadhu, Anil Boda, Nidhi Tiwari, Amit Das, S K Musharaf Ali, Dibyendu Bhattacharya, Badri N Pandey, Amit Kumar
ABSTRACT
During civil, nuclear or defense activities, internal contamination of actinides in humans and mitigation of their toxic impacts are of serious concern. Considering the health hazards of thorium (Th) internalization, an attempt was made to examine the potential of ten rationally-selected compounds/formulations to decorporate Th ions from physiological systems. The Th-induced hemolysis assay with human erythrocytes revealed good potential of tiron, silibin (SLB), phytic acid (PA) and Liv.52® (L52) for Th decorporation, in comparison to diethylenetriaminepentaacetic acid, an FDA-approved decorporation drug. This was further validated by decorporation experiments with relevant human cell models (erythrocytes and liver cells) and biological fluid (blood) under pre-/post-treatment conditions, using inductively coupled plasma mass spectrometry (ICP-MS) and transmission electron microscopy (TEM). Furthermore, density functional theory-based calculations and extended X-ray absorption fine structure (EXAFS) spectroscopy confirmed the formation of Th complex by these agents. Amongst the chosen biocompatible agents, tiron, SLB, PA and L52 hold promise to enhance Th decorporation for human application.