MilliporeSigma
  • Efficient Enhancement of Electron Transport and Collection Capability in PTB7:PC71 BM-based Solar Cells Enabled by Sulforhodamine Cathode Interlayers.
Efficient Enhancement of Electron Transport and Collection Capability in PTB7:PC71 BM-based Solar Cells Enabled by Sulforhodamine Cathode Interlayers.
Chemistry, an Asian journal (2019-02-26)
Wei Song, Wang Li, Ruixiang Peng, Billy Fanady, Jiaming Huang, Wenqing Zhu, Lingchao Xie, Tao Lei, Ziyi Ge
ABSTRACT
Development of low-cost water-/alcohol-soluble interfacial materials is a crucial issue to promote the commercialization of polymer solar cells (PSCs). Herein, two derivatives of low-cost rhodamine, called sulforhodamine 101 (SR101) and sulforhodamine B (SRB), are applied as cathode interfacial layers (CILs) to effectively improve the charge-carrier transportation and collection, reduce the work function (WF) of Al counter electrode, and decrease the series resistance and charge recombination in the PSCs. As a result, SR101-based devices show excellent performance with the highest power conversion efficiency (PCE) of 9.10 %, superior to that of both the control devices with MeOH/Al and Ca/Al. Notably, sulforhodamine is commercially available with low cost and great solution-processability. This work demonstrates that sulforhodamine has a great potential as a CIL material,which is suitable for the large-area fabrication process and commercialization of highly efficient PSCs.