MilliporeSigma
  • MiR-19b-3p attenuates IL-1β induced extracellular matrix degradation and inflammatory injury in chondrocytes by targeting GRK6.
MiR-19b-3p attenuates IL-1β induced extracellular matrix degradation and inflammatory injury in chondrocytes by targeting GRK6.
Molecular and cellular biochemistry (2019-06-23)
Liang Duan, Dapeng Duan, Wenbo Wei, Zhengming Sun, Honghai Xu, Lipeng Guo, Xuanhui Wu
ABSTRACT
Osteoarthritis (OA) is characterized by degradation of articular cartilage. MiRNAs are involved in the regulation of chondrogenesis and OA. We aimed to investigate effects and mechanisms of miR-19b-3p in regulating chondrocytes viability, cartilage degradation and inflammatory response. Primary chondrocytes were isolated from cartilages in control subjects and patients with OA. Murine ATDC5 cells were pre-conditioned with IL-1β in vitro. Expressions and interaction of miR-19b-3p with G protein-coupled receptor kinase 6 (GRK6), and their effects on inflammation, chondrocytes viability and cartilage degradation were determined after miR-19b-3p mimic or GRK6 siRNA transfection. MiR-19b-3p was significantly decreased in OA chondrocytes and IL-1β-stimulated ATDC5 cells, in paralleled with the elevated type-II-collagen, aggrecan, MMP13 and GRK6 expression. MiR-19b-3p mimic dramatically increased the viability of chondrocytes and suppressed cell apoptosis. It also increased type-II-collagen, aggrecan expression and glycosaminoglycan (sGAG) content, and decreased the expression of MMP-1 and MMP-13 that controlled by IL-1β. Overexpression of miR-19b-3p inhibited the production of IL-6 and IL-8 in ATDC5 cells. However, the protective effects of miR-19b-3p mimic on IL-1β induced cell death; IL-8 production and sGAG decrease were greatly discounted by GRK6 lentiviral vectors. Luciferase reporter assay confirmed that GRK6 gene was a direct target ofmiR-19b-3p. GRK6 siRNA transfection antagonized the IL-1β-induced chondrocytes injury, extracellular matrix degradation and inflammatory response. MiR-19b-3p mimic and GRK6 siRNA showed comparable inhibitory effect on IL-1β-provoked NF-κB as reflected by the expression of p-p65. NF-κB translocation inhibition with PS1154 reversed the effects of IL-1β on IL-8 and sGAG. Collectively, miR-19b-3p attenuated OA by targeting GRK6-NF-κB pathway.