MilliporeSigma
  • The Role of Polyethylene Glycol Size in Chemical Spectra, Cytotoxicity, and Release of PEGylated Nanoliposomal Cisplatin.
The Role of Polyethylene Glycol Size in Chemical Spectra, Cytotoxicity, and Release of PEGylated Nanoliposomal Cisplatin.
Assay and drug development technologies (2019-05-14)
Masoomeh Shirzad, Saeed Jamehbozorgi, Iman Akbarzadeh, Hamid Reza Aghabozorg, Abbas Amini
ABSTRACT
This study aimed to synthesize methoxy polyethylene glycol propionaldehyde (mPEG20,000-ALD) for the preparation of PEGylated nanoliposomal cisplatin. Nanocarriers such as liposomes are developed for a wide range of drug delivery systems. PEG with high molecular weight (Mw) is used to coat the liposomes. In this study, simulated Fourier transform infrared (FTIR) spectra of mPEG-ALD were obtained using density functional theory (DFT) calculations and then compared with actual FTIR spectrum of mPEG20,000-ALD (Mw = 20 kDa). We found that the intensity of C = O stretching vibration at 1,700 cm-1 related to the carbonyl functional group of mPEG20,000-ALD was very weak. The results of DFT calculations of mPEG-ALD showed that by increasing the Mw of mPEG-ALD, the intensity of C = O stretching vibration related to the carbonyl functional group of mPEG-ALD was decreased, so we concluded the hypothesis of decreasing the intensity of C = O stretching vibration at 1,700 cm-1 as a result of increasing the Mw of mPEG-ALD. In vitro release of cisplatin showed that the percentages of released cisplatin from PEGylated nanoliposomal cisplatin and free cisplatin were determined to be 46 ± 2% and 97 ± 3% after 35 h, respectively. Cytotoxicity of free cisplatin and PEGylated nanoliposomal cisplatin was evaluated and related half-maximal inhibitory concentration on human ovarian cancer cell line A2780CP was obtained to be 76.6 ± 3.1 and 46.6 ± 2.3 μg/mL, respectively.