MilliporeSigma
Acyclovir inhibits Cyprinid herpesvirus 3 multiplication in vitro.
Journal of fish diseases (2018-08-26)
Agnieszka Troszok, Ludmiła Kolek, Joanna Szczygieł, Joanna Wawrzeczko, Ewa Borzym, Michał Reichert, Teresa Kamińska, Tomasz Ostrowski, Patrycja Jurecka, Mikołaj Adamek, Krzysztof Rakus, Ilgiz Irnazarow
ABSTRACT
Cyprinid herpesvirus 3 (CyHV-3), also known as koi herpesvirus (KHV), is an aetiological agent of a virulent and lethal disease in common and koi carp. In this study, we examined in vitro the anti-CyHV-3 activity of acyclovir (ACV), nucleoside analogue commonly used against human herpesviruses, as well as acyclovir monophospate (ACV-MP). The cytotoxicity of the ACV and the ACV-MP for two common carp cell lines, CCB (Common carp brain) and KF1 (Koi carp fin 1), was determined by means of MTT and crystal violet assays. In subsequent studies, the concentration of 66.67 μM was applied. The ACV and the ACV-MP (66.67 μM) inhibited a cytopathic effect (CPE) induced by the CyHV-3 virus in the CCB (ACV by 66%, ACV-MP by 58%) and the KF1 (ACV by 25%, ACV-MP by 37%). The viral load measured by the means of TaqMan qPCR was reduced in a range of 67%-93% depending on the analogue, the cell line and the time of incubation. The expression of viral genes (ORF149, ORF3, ORF134 and ORF78) in CCB cells infected with the CyHV-3 was strongly downregulated within the range of 78%-91%. In summary, both the ACV and the ACV-MP can inhibit CyHV-3 replication in vitro.