MilliporeSigma
  • Complex formation equilibria of polyamine ligands with copper(II) and zinc(II).
Complex formation equilibria of polyamine ligands with copper(II) and zinc(II).
Journal of inorganic biochemistry (2019-02-27)
Valeria M Nurchi, Guido Crisponi, Gavino Sanna, Inmaculada Pérez-Toro, Juan Niclos-Gutierrez, Maria Josefa Gonzalez-Perez, Alicia Domínguez Martín
ABSTRACT
A comprehensive study of the protonation equilibria of a series of polyamine ligands along with their complex formation equilibria with Cu2+ and Zn2+ is reported in this work. The primary aim of this study has been the achievement of homogeneous thermodynamic data on these ligands, in order to evaluate their influence on the homeostatic equilibria of essential metal ions (Cu2+ and Zn2+) in biological fluids. These polyamines are largely used as linkers in the building of chelating agents for iron overload. Potentiometric and spectrophotometric techniques were used for the characterization of protonation and complex formation constants. In addition, the characterization of the formed complexes is discussed together with selected solid-state crystal structures, remarking the influence of the length of the chain and of the linear or tetradentate tripod nature of the polyamine ligands on the stability of the complexes.