MilliporeSigma
Fos-like antigen 2 (FOSL2) promotes metastasis in colon cancer.
Experimental cell research (2018-08-17)
Shuo Li, Xue-Dong Fang, Xiu-Ying Wang, Bing-Yuan Fei
ABSTRACT
Among different cancers, incidence and mortality of colorectal cancer (CRC) is one of the highest. KRAS mutation is one of the underlying features in the pathogenesis of CRC with CRC tumors harboring mutant KRAS exhibiting a more aggressive behavior compared to CRC tumors with wild type KRAS. We had earlier shown that the microRNA-143 (miR-143) replenishment not only chemosensitizers CRC cell line with mutant KRAS instead of wild-type KRAS gene, to paclitaxel-mediated cytotoxicity, but also inhibits cell migration and invasion ability. Hence, the study aimed to determine how miR-143 replenishment is inhibiting pre-metastatic behavior in CRC cells with mutant KRAS. Top ten mRNA targets of miR-143 as predicted by TargetScan were evaluated by qRT-PCR in LoVo cells which were performed mock transfection or miR-143 mimic transfection. Evaluation of the changes in cognate mRNA target(s) was done in 30 paired CRC tissue and tumor adjacent normal tissue specimens and in LoVo cells by western blot. Effect of the mRNA target on pro-metastatic behavior was assayed by gain- and loss-of-function studies using a combination of western blotting and in vitro cell proliferation and transwell migration/invasion assay in LoVo cells and in the normal colonic epithelium cell line FHC. In vivo effect of the cognate mRNA target on CRC metastasis was assayed by xenograft assay. Of the 10 predicted mRNA targets, FOSL2 (P < 0.05) and IGFBP5 (P > 0.05) was down regulated in LoVo cells transfected with the miR-143 mimic. FOSL2 mRNA levels were significantly downregulated in CRC tissue specimens compared with adjacent normal tissue (P < 0.05). Immunoblot analysis showed that FOSL2, but not IGFBP5, protein expression is down regulated in LoVo cells after the miR-143 mimic transfection. FOSL2 overexpression in the normal colonic epithelial cell line FHC or siRNA-mediated silencing in LoVo cells induced and repressed, respectively, pro-mesenchymal cell features. Whereas manipulation of FOSL2 expression did not have any effect on cell proliferation rates, silencing its expression inhibited cell migration and invasion ability in vitro. In addition, silencing of FOSL2 expression in the LoVo cells can significantly inhibited invasion of hepatic, while no effect was found for tumorigenic potential. Our results suggest that FOSL2 is a critical regulator of CRC metastasis and might be an important marker for prognostic in CRC patients.