MilliporeSigma
  • Advanced method for fabrication of molecularly imprinted mesoporous organosilica with highly sensitive and selective recognition of glyphosate.
Advanced method for fabrication of molecularly imprinted mesoporous organosilica with highly sensitive and selective recognition of glyphosate.
Scientific reports (2019-07-18)
Youngdo Kim, Jaeho Lee, Ik-Soo Shin
ABSTRACT
In this study, we synthesized molecularly imprinted mesoporous organosilica (MIMO) in the presence of a new precursor having a zwitterionic functional group and an imprint molecule, namely, glyphosate (MIMO-z). The precursor-glyphosate complex engaged in a typical base-catalyzed sol-gel reaction and the introduced zwitterion group remained intact in the framework after the extraction process had been completed. To test the rebinding performance of the target molecule, graphene quantum dots were encapsulated (MIMO-zQ) into pores and the fluorescence intensity change was monitored according to the concentration of glyphosate. When the MIMO-zQ suspension was diluted into the glyphosate solutions, notable fluorescence quenching occurred, right down to sub-nanomolar levels of concentration; 9.2 ± 0.18% quenching at 0.1 nM (0.017 ppb, 17 pg/mL). This result is one of the best reported to date for sensing using MIMO. The synthesized probe also exhibited a distinct signal compared to a series of competing compounds, aminomethylphosphonic acid and glycine; 4.3 ± 0.019% and 3.7 ± 0.041% quenching at 100 nM.