MilliporeSigma
  • Influence of Alcohol Consumption on Body Mass Gain and Liver Antioxidant Defense in Adolescent Growing Male Rats.
Influence of Alcohol Consumption on Body Mass Gain and Liver Antioxidant Defense in Adolescent Growing Male Rats.
International journal of environmental research and public health (2019-07-03)
Aleksandra Kołota, Dominika Głąbska, Michał Oczkowski, Joanna Gromadzka-Ostrowska
ABSTRACT
The World Health Organization (WHO) reported that alcohol consumption is a serious problem in adolescents. The aim of the study was to assess the influence of the time of exposure of various alcoholic beverages on body mass as well as on select parameters of liver antioxidant defense in adolescent Wistar rats. Thirty-day-old animals were divided into 12 groups (six animals in each): control and groups receiving various beverages containing 10% of alcohol (ethanol, red wine, beer), observed for two, four, and six weeks. The body weight gain and energy supply were analyzed for body mass assessment. The catalase (CAT), superoxide dismutase (SOD), glutathione peroxidase, transferase (GST), reductase activities, total antioxidant status, and glutathione level (GSH) were analyzed, for a liver antioxidant defense assessment. Group receiving red wine was characterized by the highest alcohol intake, lowest dietary intake, and highest total energy supply (p < 0.05). However, this did not influence body weight gain (p > 0.05). Reduced diet intake in groups receiving alcohol was counterbalanced by its energy value. Therefore, the energy supply was not lower than for the control (p > 0.05). Alcohol consumption and the experiment duration influenced CAT, SOD, and GST activities and GSH level. Alcohol consumption may influence hepatic antioxidant defense in adolescent male rats, but without influence on body weight gain.