MilliporeSigma
  • Maternal Consumption of a Diet Rich in Maillard Reaction Products Accelerates Neurodevelopment in F1 and Sex-Dependently Affects Behavioral Phenotype in F2 Rat Offspring.
Maternal Consumption of a Diet Rich in Maillard Reaction Products Accelerates Neurodevelopment in F1 and Sex-Dependently Affects Behavioral Phenotype in F2 Rat Offspring.
Foods (Basel, Switzerland) (2019-05-22)
Melinda Csongová, Emese Renczés, Veronika Šarayová, Lucia Mihalovičová, Jakub Janko, Radana Gurecká, Antonio Dario Troise, Paola Vitaglione, Katarína Šebeková
ABSTRACT
Thermal processing of foods at temperatures > 100 °C introduces considerable amounts of advanced glycation end-products (AGEs) into the diet. Maternal dietary exposure might affect the offspring early development and behavioral phenotype in later life. In a rat model, we examined the influence of maternal (F0) dietary challenge with AGEs-rich diet (AGE-RD) during puberty, pregnancy and lactation on early development, a manifestation of physiological reflexes, and behavioral phenotype of F1 and F2 offspring. Mean postnatal day of auditory conduit and eye opening, or incisor eruption was not affected by F0 diet significantly. F1 AGE-RD offspring outperformed their control counterparts in hind limb placing, in grasp tests and surface righting; grandsons of AGE-RD dams outperformed their control counterparts in hind limb placing and granddaughters in surface righting. In a Morris water maze, female AGE-RD F1 and F2 offspring presented better working memory compared with a control group of female offspring. Furthermore, male F2 AGE-RD offspring manifested anxiolysis-like behavior in a light dark test. Mean grooming time in response to sucrose splash did not differ between dietary groups. Our findings indicate that long-term maternal intake of AGE-RD intergenerationally and sex-specifically affects development and behavioral traits of offspring which have never come into direct contact with AGE-RD.